Order Entry
Sweden
ContactUsLinkComponent
EDIL3 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human EDIL3 in cell culture su pernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5150
Supplier:  Abnova
EDIL3 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human EDIL3 in cell culture su pernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5150
Supplier:  Abnova