0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Pspn (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of mouse persephin in cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
ABNOKA5149
:
Abnova
Pspn (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of mouse persephin in cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
ABNOKA5149
:
Abnova