0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
COLUMN EC NUCLEOSIL 100-5 C18 200X4.6MM 1 * 1 items
Catalog # MANA720006.46
Supplier:
MACHEREY-NAGEL
Expand image
COLUMN EC NUCLEOSIL 100-5 C18 200X4.6MM 1 * 1 items
Catalog # MANA720006.46
Supplier:
MACHEREY-NAGEL