Order Entry
Sweden
ContactUsLinkComponent
COMT (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human COMT in cell culture supe rnates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5100
Supplier:  Abnova
COMT (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human COMT in cell culture supe rnates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5100
Supplier:  Abnova