Order Entry
Sweden
ContactUsLinkComponent
Cxcl2 (Rat) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of rat MIP-2 in cell culture supern ates, serum and plasma (heparin, EDTA). 1 * 1 KIT
  ABNOKA5062
 :  Abnova
Cxcl2 (Rat) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of rat MIP-2 in cell culture supern ates, serum and plasma (heparin, EDTA). 1 * 1 KIT
  ABNOKA5062
 :  Abnova