Order Entry
Sweden
ContactUsLinkComponent
Ctf1 (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of mouse CT-1 in cell culture supe rnates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5056
Supplier:  Abnova
Ctf1 (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of mouse CT-1 in cell culture supe rnates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5056
Supplier:  Abnova