Order Entry
Sweden
ContactUsLinkComponent
Human ErbB3, His Tag (ER3-H5223) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession P21860-1). 1 * 100 µG
Catalog # ACRBER3-H5223100UG
Supplier:  ACRO Biosystems
Human ErbB3, His Tag (ER3-H5223) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession P21860-1). 1 * 100 µG
Catalog # ACRBER3-H5223100UG
Supplier:  ACRO Biosystems
Frequently Bought Together