Order Entry
Sweden
ContactUsLinkComponent
Capillary, coated, Zero Flow brand, covalently bonded, neutral, no flow 50um ID by 1 Meter (Nominal Length). Use in Capillary Electrophoresis. 1/pack. 1 * 1 items
Catalog # MICS04650-ZF
Leverantör:  MICROSOLV
undefined
Capillary, coated, Zero Flow brand, covalently bonded, neutral, no flow 50um ID by 1 Meter (Nominal Length). Use in Capillary Electrophoresis. 1/pack. 1 * 1 items
Catalog # MICS04650-ZF
Leverantör:  MICROSOLV
Frequently Bought Together