Polyethylene glycol monomethyl ether 2000
500 g
TCIAP2186-500G
: TCI
: Methoxypolyethylene glycols, Methoxy poly(ethylene glycol), PEG MME, Poly(ethylene glycol) methyl ether, mPEG, Methoxypolyethylene glycols-average M.W. 1000, m-PEG-OH
- :9004-74-4
- :CH₃(OCH₂CH₂)nOH
- :MFCD00084416
- Appearance:White to Almost white powder to crystaline
- Average molecular weight:1,900 to 2,100
- Hydroxyl Value:25.0 to 30.0
- Melting point:49.0 to 55.0 deg-C
- Förp.:500 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



