Order Entry
Sweden
ContactUsLinkComponent
Bimetal dial therm., axial, 0+120:2°C, built-in length 45mm, diameter of case 63mm, thread R 1/2 1 * 1 items
Catalog # AMART110304
Supplier:  Amarell
Bimetal dial therm., axial, 0+120:2°C, built-in length 45mm, diameter of case 63mm, thread R 1/2 1 * 1 items
Catalog # AMART110304
Supplier:  Amarell