Order Entry
Sweden
ContactUsLinkComponent
Therm. to set Amarell Universal, +197+252:0,1°C, 400x9mm, enclosed scale, cap.lens front, colourless, gallium filled, total immersio n, richter-top finish, suitable for government calibration 1 * 1 items
  AMARN71014-GA
 :  Amarell
Therm. to set Amarell Universal, +197+252:0,1°C, 400x9mm, enclosed scale, cap.lens front, colourless, gallium filled, total immersio n, richter-top finish, suitable for government calibration 1 * 1 items
  AMARN71014-GA
 :  Amarell