Order Entry
Sweden
ContactUsLinkComponent
Range finders, DIN 12793, 0,60-0,85:0,005g/cm , ref. temp. 20C, without therm., 240mm +/-15mm long 1 * 1 items
Catalog # AMARH820480
Supplier:  Amarell
Range finders, DIN 12793, 0,60-0,85:0,005g/cm , ref. temp. 20C, without therm., 240mm +/-15mm long 1 * 1 items
Catalog # AMARH820480
Supplier:  Amarell