Order Entry
Sweden
ContactUsLinkComponent
API Gravity hydrometer ASTM, 29H-62, 40+46:0,1API, ref. temp. 60F, without therm., 163mm long 1 * 1 items
Katalog # AMARH825136
Leverantör:  Amarell
API Gravity hydrometer ASTM, 29H-62, 40+46:0,1API, ref. temp. 60F, without therm., 163mm long 1 * 1 items
Katalog # AMARH825136
Leverantör:  Amarell