Thermometer similar to IP 73 C, solid stem, white backed, -5+400:1°C, gallium filled, durable pigment, suitable for government verif ication 1 * 1 items
Thermometer similar to IP 73 C, solid stem, white backed, -5+400:1°C, gallium filled, durable pigment, suitable for government verif ication 1 * 1 items