Order Entry
Sweden
ContactUsLinkComponent
Hydrometer DIN 12791/BS 718, M50, 1,70-1,75:0,0010g/cm , ref. temp. 20C, without therm., max. 270mm long, suitable for government v erification 1 * 1 items
Catalog # AMARH808044
Supplier:  Amarell
Hydrometer DIN 12791/BS 718, M50, 1,70-1,75:0,0010g/cm , ref. temp. 20C, without therm., max. 270mm long, suitable for government v erification 1 * 1 items
Catalog # AMARH808044
Supplier:  Amarell