Order Entry
Sweden
ContactUsLinkComponent
Thermometer similar to IP 51 C, solid stem, white backed, 35+120:0,5C, red special liquid, durable pigment, suitable for government calibration 1 * 1 items
  AMARA340740-FL
 :  Amarell
Thermometer similar to IP 51 C, solid stem, white backed, 35+120:0,5C, red special liquid, durable pigment, suitable for government calibration 1 * 1 items
  AMARA340740-FL
 :  Amarell