Order Entry
Sweden
ContactUsLinkComponent
Thermometer similar to IP 48 C, solid stem, white backed, -38+30:0,5°C, red special liquid, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARA340680-FL
Supplier:  Amarell
Thermometer similar to IP 48 C, solid stem, white backed, -38+30:0,5°C, red special liquid, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARA340680-FL
Supplier:  Amarell