Order Entry
Sweden
ContactUsLinkComponent
Density hydrometer, 1,180-1,240:0,001g/cm , ref. temp. 20C, with therm., 350mm long, special liquid 1 * 1 items
Katalog # AMARH800260-FL
Supplier:  Amarell
Density hydrometer, 1,180-1,240:0,001g/cm , ref. temp. 20C, with therm., 350mm long, special liquid 1 * 1 items
Katalog # AMARH800260-FL
Supplier:  Amarell