Order Entry
Sweden
ContactUsLinkComponent
Distillation therm., similar to DIN, 0+150:1°C, enclosed scale, cap. prismatic, colourless, red special liquid, total immersion, len gth 260mm, pin-top finish with ring 1 * 1 items
Catalog # AMARD259606-FL
Supplier:  Amarell
Distillation therm., similar to DIN, 0+150:1°C, enclosed scale, cap. prismatic, colourless, red special liquid, total immersion, len gth 260mm, pin-top finish with ring 1 * 1 items
Catalog # AMARD259606-FL
Supplier:  Amarell