Order Entry
Sweden
ContactUsLinkComponent
Thermometer similar to BS 1365, SB105C/total, 95+105:0,1°C, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liquid, 235mm long, total immersion, durable pigment, null
Catalog # AMARA402562-CB
Supplier:  Amarell
Thermometer similar to BS 1365, SB105C/total, 95+105:0,1°C, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liquid, 235mm long, total immersion, durable pigment, null
Catalog # AMARA402562-CB
Supplier:  Amarell