0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Hydrometer Baume, 40-60:0,1B, without therm., ref. temp. 15C, 330mm long 1 * 1 items
AMARH842884
:
Amarell
Hydrometer Baume, 40-60:0,1B, without therm., ref. temp. 15C, 330mm long 1 * 1 items
AMARH842884
:
Amarell