Order Entry
Sweden
ContactUsLinkComponent
Thermometer similar to BS 1704, F/75, 0+250:1C, solid stem, white backed, blue special liquid, 75mm immersion, 300mm long, durable pigment 1 * 1 items
Catalog # AMARA402110-FL
Supplier:  Amarell
Thermometer similar to BS 1704, F/75, 0+250:1C, solid stem, white backed, blue special liquid, 75mm immersion, 300mm long, durable pigment 1 * 1 items
Catalog # AMARA402110-FL
Supplier:  Amarell