Order Entry
Sweden
ContactUsLinkComponent
Thermometer similar to BS 1704:1985, F/total, 0+250:1°C, solid stem, white backed, blue special liquid, total immersion, 300mm long, durable pigment 1 * 1 items
Catalog # AMARA402090-FL
Supplier:  Amarell
Thermometer similar to BS 1704:1985, F/total, 0+250:1°C, solid stem, white backed, blue special liquid, total immersion, 300mm long, durable pigment 1 * 1 items
Catalog # AMARA402090-FL
Supplier:  Amarell