Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

3'-Nitropropiophenone

Bottle, 5 g
Catalog # APOSOR11232-5G
Irritant
Supplier:   Apollo Scientific
3'-Nitropropiophenone
Catalog # APOSOR11232-5G
Supplier:  Apollo Scientific
CAS Number:  17408-16-1
3'-Nitropropiophenone

Bottle, 5 g

CAS Number:  17408-16-1
Price
1 375,00 kr
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  m-Nitropropiophenone, 1-(3-Nitrophenyl)propan-1-one
  • CAS Number:17408-16-1
  • Formula:C₉H₉NO₃
  • MDL Number:MFCD00051516
  • Molecular Weight:179.18 g/mol
  • Melting Pt:99…102 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.