Potassium iodide
Bottle, 250 g
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: Dipotassium diiodide, Kali iodide
- CAS Number:7681-11-0
- Formula:Kl
- MDL Number:MFCD00011405
- Molecular Weight:166 g/mol
- Boiling Pt:1330 °C
- Melting Pt:681 °C
- Density:3.13 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:231-659-4
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Bottle
- Pk:250 g



