Order Entry
Sweden
Orders LinkContactUsLinkComponent
L(+)-Valine ≥99.5%
Bottle, 1 kg
  APOSBIV0722-1KG
L(+)-Valine ≥99.5%
  APOSBIV0722-1KG
 :  72-18-4
L(+)-Valine ≥99.5%
Bottle, 1 kg
 :  72-18-4
615,00 kr
1 * 1 kg
USP, pyrogen free
 :  H-Val-OH, Valine, (S)-(+)-Valine
  •  :72-18-4
  •  :C₅H₁₁NO₂
  •  :MFCD00064220
  •  :117.15 g/mol
  •  :295…300 °C
  •  :Ambient
  •  :200-773-6
  • Pack type:
    Bottle
  • Pk:
    1 kg