Order Entry
Sweden
Orders LinkContactUsLinkComponent
L(+)-Valine ≥99.5%

L(+)-Valine ≥99.5%

Bottle, 100 g
Catalog # APOSBIV0722-100G
Irritant
Supplier:   Apollo Scientific
L(+)-Valine ≥99.5%
Catalog # APOSBIV0722-100G
Supplier:  Apollo Scientific
CAS Number:  72-18-4
L(+)-Valine ≥99.5%

Bottle, 100 g

CAS Number:  72-18-4
Price
260,00 kr
Log in for availability
1 * 100 g
Product Details & Documents

USP, pyrogen free

Synonyms:  H-Val-OH, Valine, (S)-(+)-Valine
  • CAS Number:72-18-4
  • Formula:C₅H₁₁NO₂
  • MDL Number:MFCD00064220
  • Molecular Weight:117.15 g/mol
  • Melting Pt:295…300 °C
  • Storage Temperature:Ambient
  • EINECS:200-773-6
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.