Order Entry
Sweden
Orders LinkContactUsLinkComponent
Recombinant Hepatitis A Virus P2C

Recombinant Hepatitis A Virus P2C

1 mg
Catalog # APOSBITP1814-1MG
Supplier:   Apollo Scientific
Recombinant Hepatitis A Virus P2C
Catalog # APOSBITP1814-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Hepatitis A Virus P2C

1 mg

CAS Number:  
Price
16 675,00 kr
Log in for availability
1 * 1 mg
Product Details & Documents
Specifications

Specifications

  • Pk:
    1 mg
  • Cat. No.:
    APOSBITP1814-1MG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    Hepatitis A Virus P2C
  • Protein synonyms:
    HAV P2C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.