Order Entry
Sweden
Orders LinkContactUsLinkComponent
D-(-)-Aspartic acid

D-(-)-Aspartic acid

Bottle, 1 kg
Catalog # APOSBIA0866-1KG
Irritant
Supplier:   Apollo Scientific
D-(-)-Aspartic acid
Catalog # APOSBIA0866-1KG
Supplier:  Apollo Scientific
CAS Number:  1783-96-6
D-(-)-Aspartic acid

Bottle, 1 kg

CAS Number:  1783-96-6
Price
1 325,00 kr
Log in for availability
1 * 1 kg
Product Details & Documents

Chiral intermediate for pharmaceuticals and agrochemicals; Materials for peptides.

Synonyms:  D-Aminosuccinic acid, D(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-Aspartic acid, (R)-()-Aminosuccinic acid, (R)-Aminosuccinic acid, (R)-2-Aminosuccinic acid, D-Asparaginic acid, D-Aspartic acid, H-D-Asp-OH
  • CAS Number:1783-96-6
  • Formula:C₄H₇NO₄
  • MDL Number:MFCD00063081
  • Molecular Weight:133.1 g/mol
  • Melting Pt:>300 °C
  • Melting Pt:>300 °C
  • Density:1.66 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:217-234-6
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.