D-(-)-Aspartic acid
Bottle, 100 g
Catalog # APOSBIA0866-100G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Chiral intermediate for pharmaceuticals and agrochemicals; Materials for peptides.
Synonyms: D-Aminosuccinic acid, D(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-Aspartic acid, (R)-()-Aminosuccinic acid, (R)-Aminosuccinic acid, (R)-2-Aminosuccinic acid, D-Asparaginic acid, D-Aspartic acid, H-D-Asp-OH
- CAS Number:1783-96-6
- Formula:C₄H₇NO₄
- MDL Number:MFCD00063081
- Molecular Weight:133.1 g/mol
- Melting Pt:>300 °C
- Melting Pt:>300 °C
- Density:1.66 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:217-234-6
Specifications
Specifications
- Pack type:Bottle
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



