Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
HANDLE SS DIAM 31 MM 1025 MM RED 1 * 1 items
Catalog # BSNC29834
Supplier:
Basan
Expand image
HANDLE SS DIAM 31 MM 1025 MM RED 1 * 1 items
Catalog # BSNC29834
Supplier:
Basan