Order Entry
Sweden
ContactUsLinkComponent
SPR sensorchip. Biacore Series S compatible. Coating: 500 nm NTA derivatized carboxymethyldextran hydrogel, 0,25 x low density 1 * 1 Items
Katalog # IBSASCBSNID500LLL
Supplier:  IBS Stock Items
SPR sensorchip. Biacore Series S compatible. Coating: 500 nm NTA derivatized carboxymethyldextran hydrogel, 0,25 x low density 1 * 1 Items
Katalog # IBSASCBSNID500LLL
Supplier:  IBS Stock Items
Frequently Bought Together