Order Entry
Sweden
ContactUsLinkComponent
SPR sensorchip. Biacore Series S compatible. Coating: 500 nm NTA derivatized carboxymethyldextran hydrogel, 0,5 x low density. 1 * 1 Items
Catalog # IBSASCBSNID500LL
Supplier:  IBS Stock Items
SPR sensorchip. Biacore Series S compatible. Coating: 500 nm NTA derivatized carboxymethyldextran hydrogel, 0,5 x low density. 1 * 1 Items
Catalog # IBSASCBSNID500LL
Supplier:  IBS Stock Items
Frequently Bought Together