Niclosamide 97+%
Glass bottle, 100 g
Catalog # ACRO461841000
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:50-65-7
- Formula:C₁₃H₈Cl₂N₂O₄
- MDL Number:MFCD00057597
- Molecular Weight:327.12 g/mol
- Melting Pt:225…230 °C
- Storage Temperature:Ambient
- UN:3077
- ADR:9,III
Specifications
Specification Test Results
- Appearance:Off-white to yellow Powder
- Infrared spectrum:Conforms
- Melting point:227 to 232 °C
- Loss on drying:≤2 % (105 °C, 4 h)
- HPLC:≥97.0 %
Specifications
- Pack type:Glass bottle
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



