0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
C-FLEX ULTRA PUMP TUBING, 0.500 ID, 0.750 OD, 0.125 WALL, LENGTH 50 FEET, 3 COILS 1 * 3 items
Catalog # GESS700-500-4
Supplier:
Saint Gobain Life Sciences
Expand image
C-FLEX ULTRA PUMP TUBING, 0.500 ID, 0.750 OD, 0.125 WALL, LENGTH 50 FEET, 3 COILS 1 * 3 items
Catalog # GESS700-500-4
Supplier:
Saint Gobain Life Sciences