Order Entry
Sweden
ContactUsLinkComponent
Cynomolgus CD40, His Tag (CD0-C52H6) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession G7PG38). 1 * 1 mg
Catalog # ACRBCD0-C52H6-1MG
Supplier:  ACRO Biosystems
Cynomolgus CD40, His Tag (CD0-C52H6) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession G7PG38). 1 * 1 mg
Catalog # ACRBCD0-C52H6-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together