Shelving systems, direct wall mounts
Shelf, single, 360 mm, 0,9 kg
Catalog # METO1WD14S
Supplier: Metro International
Jump to:
Product Details & Documents
Direct wall mount brackets allow the installation of wire or solid shelves directly onto a wall surface. Each brackets consists of one shelf support and mounting plate, with two shelf collar caps. Use single support at shelf ends and double support for adjoining shelves.
Specifications
- Description:Shelf, single, 360 mm
- Seq:02
- Weight:0,9 kg
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



