Order Entry
Sweden
ContactUsLinkComponent
CALIBRATED NOMINALLY 50% REFLECTIVE SINTERED PTFE POLYMER DISC 8 MM THICK X 58 MM DIAMETER WITHIN A SEALABLE CONTAINER SUITABLE FOR THE NIRA II 60 MM SPINNER. 1 * 1 items
Catalog # PERCL1281924
Supplier:  PerkinElmer
CALIBRATED NOMINALLY 50% REFLECTIVE SINTERED PTFE POLYMER DISC 8 MM THICK X 58 MM DIAMETER WITHIN A SEALABLE CONTAINER SUITABLE FOR THE NIRA II 60 MM SPINNER. 1 * 1 items
Catalog # PERCL1281924
Supplier:  PerkinElmer