THE CINNABAR SPRAY CHAMBER IS A MINI-CYCLONIC SPRAY CHAMBER WHICH GIVES GOOD SENSITIVITY AT VERY LOW UPTAKES. THE CINNABAR'S DESIGN AND LOW VOLUME IS IDEAL FOR THE REDUCTION OF MEMORY AND MATRIX EFFECTS AND VERY FAST WASHOUT. MATERIAL: BOROSILICATE G 1 * 1 items
Katalog # PERCN8121063
Leverantör: PerkinElmer
Frequently Bought Together