Drop-n-Stain™ CF543 goat anti-rabbit IgG (H+L) is provided in a convenient dropper bottle for easy dispensing without pipetting. To minimize cross-reactivity, the antibody has been adsorbed against human, mouse, and rat serum. 1 * 5 mL
Catalog # BTIU20968
Supplier: Biotium
Frequently Bought Together