HYDROCHLORIC ACID STD SOLUTION 0.1N 1 * 1 L
Catalog # HACH1481253
Supplier: Hach
Frequently Bought Together
Iodine 0.05 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Dimethyl sulphoxide ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals