acd (acid citrate dextrose) solution a andacd (acid citrate dextrose)solution b are anticoagulants for whole blood and also act as a cidifying agents to platelet poor plasma. 1 * 100 mL
Catalog # GENO786-820
Supplier: G-Biosciences
Frequently Bought Together