Order Entry
Sweden
ContactUsLinkComponent
mops sds buffer kit is a complete kit to resolve small- to medium-sized proteins under denaturing conditions on bis-tris gels. 1 * 1 KIT
Catalog # GENO786-1218
Supplier:  G-Biosciences
mops sds buffer kit is a complete kit to resolve small- to medium-sized proteins under denaturing conditions on bis-tris gels. 1 * 1 KIT
Catalog # GENO786-1218
Supplier:  G-Biosciences