education: g-biosciences, chymosin, recombinant rennin, 100ml. biochemistry classroom kits 1 * 100 mL
Catalog # GENOBTNM-0025
Supplier: G-Biosciences
education: g-biosciences, chymosin, recombinant rennin, 100ml. biochemistry classroom kits 1 * 100 mL
Catalog # GENOBTNM-0025
Supplier: G-Biosciences
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Cell culture flasks, Corning®
Supplier: Corning
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Supplier: Dr. Weigert
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
ALPHA-AMYLASE 1 * 25 g
Supplier: G-Biosciences
LAMP UV 30 W ADVANCE 3, 4, 6 1 * 1 items
Supplier: Azbil Telstar Technologies
Turbidity calibration standards, formazin, Supelco®
Supplier: Merck