Order Entry
Sweden
ContactUsLinkComponent
Monoclonal anti-GFP is designed to detect enhanced GFP and GFP containing recombinant proteins. This product is prepared by labeling high quality monoclonal mouse anti-GFP IgG (H+L) with with the far red fluorescent dye CF™660R. 1 * 100 µl
Catalog # BTIU20481
Supplier:  Biotium
Monoclonal anti-GFP is designed to detect enhanced GFP and GFP containing recombinant proteins. This product is prepared by labeling high quality monoclonal mouse anti-GFP IgG (H+L) with with the far red fluorescent dye CF™660R. 1 * 100 µl
Catalog # BTIU20481
Supplier:  Biotium