Order Entry
Sweden
ContactUsLinkComponent
SH2D4A (SH2 domain containing protein 4A), also known as SH2A, is a ubiquitously expressed 454 amino acid docking protein that belon gs to the SH2 signaling protein family. 1 * 100 µG
  BWRLBS61270
SH2D4A (SH2 domain containing protein 4A), also known as SH2A, is a ubiquitously expressed 454 amino acid docking protein that belon gs to the SH2 signaling protein family. 1 * 100 µG
  BWRLBS61270
Frequently Bought Together