Order Entry
Sweden
ContactUsLinkComponent
PHACS (putative human ACS), also known as ACC or ACS (1-aminocyclopropane-1-carboxylate synthase homolog), is a 501 amino acid prote in that belongs to the alpha family of pyridoxal-5-prime-phosphate enzymes. 1 * 100 µG
  BWRLBS8880
PHACS (putative human ACS), also known as ACC or ACS (1-aminocyclopropane-1-carboxylate synthase homolog), is a 501 amino acid prote in that belongs to the alpha family of pyridoxal-5-prime-phosphate enzymes. 1 * 100 µG
  BWRLBS8880
Frequently Bought Together