DNA replication, recombination and repair, all of which are necessary for genomic stability, require the presence of exonucleases. I n DNA replication, these enzymes are involved in the processing of Okazaki fragments, whereas in DNA repair 1 * 100 µG
Catalog # BWRLBS61176
Supplier: Bioworld Technology
Frequently Bought Together