Order Entry
Sweden
ContactUsLinkComponent
LZTR1, leucine-zipper-like transcriptional regulator 1, is a member of the BTBkelch superfamily. LZTR1 contains 2 BTB (POZ) domains and 6 Kelch repeats. 1 * 100 µG
Catalog # BWRLBS8853
LZTR1, leucine-zipper-like transcriptional regulator 1, is a member of the BTBkelch superfamily. LZTR1 contains 2 BTB (POZ) domains and 6 Kelch repeats. 1 * 100 µG
Catalog # BWRLBS8853
Frequently Bought Together