Order Entry
Sweden
ContactUsLinkComponent
The enzyme encoded by this gene is an arylesterase that mainly hydrolyzes paroxon to produce p-nitrophenol. Paroxon is an organophos phorus anticholinesterase compound that is produced in vivo by oxidation of the insecticide parathion. 1 * 100 µG
  BWRLBS8844
The enzyme encoded by this gene is an arylesterase that mainly hydrolyzes paroxon to produce p-nitrophenol. Paroxon is an organophos phorus anticholinesterase compound that is produced in vivo by oxidation of the insecticide parathion. 1 * 100 µG
  BWRLBS8844
Frequently Bought Together